Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,N-Dimethyl-1-(naphthalen-1-yl)methanamine
N,N-Dimethyl-1-(naphthalen-1-yl)methanamine 95%
N,N-Dimethyl-1-(naphthalen-1-yl)methanamine 95%
N,N-Dimethyl-1-(naphthalen-1-yl)methanamine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
N,N-Dimethyl-1-(naphthalen-1-yl)methanamine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
16413-71-1
:
Formula:
C₁₃H₁₅N
MDL Number:
MFCD05124184
:
185.265 g/mol
Storage Temperature:
Ambient