Order Entry
Sweden
ContactUsLinkComponent
2-(Furan-2-yl)phenol 98%
2-(Furan-2-yl)phenol 98%
Catalog # APOSOR954762-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Furan-2-yl)phenol 98%
Catalog # APOSOR954762-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.169 g/mol
Storage Temperature:Ambient
CAS Number:106584-14-9
Formula:C₁₀H₈O₂
MDL Number:MFCD06802855