Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Acetyl-5-hydroxytetralin
6-Acetyl-5-hydroxytetralin 95%
6-Acetyl-5-hydroxytetralin 95%
6-Acetyl-5-hydroxytetralin 95%
Leverantör:
Apollo Scientific
Certifikat
Expand image
6-Acetyl-5-hydroxytetralin 95%
Leverantör:
Apollo Scientific
Om denna produkt
CAS-nummer:
95517-07-0
:
Formel:
C₁₂H₁₄O₂
MDL-nummer:
MFCD06658289
:
190.238 g/mol
Förvaringstemperatur:
Ambient