Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Oxazolo[5,4-b]pyridine
Oxazolo[5,4-b]pyridine 95%
Oxazolo[5,4-b]pyridine 95%
Oxazolo[5,4-b]pyridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Oxazolo[5,4-b]pyridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
273-62-1
:
Formula:
C₆H₄N₂O
MDL Number:
MFCD13175184
:
120.109 g/mol
Storage Temperature:
Ambient