4-methyl-1,3-benzothiazole-2-thiol 95%
Catalog # APOSOR909266-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:181.278 g/mol
Storage Temperature:Ambient
CAS Number:2268-77-1
Formula:C₈H₇NS₂
MDL Number:MFCD11044826