(S)-tert-Butyl (piperidin-2-ylmethyl)carbamate
Catalog # APOSOR52456-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:214.305 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:139004-93-6
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD11975335