Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(Bromomethyl)cyclopropane-1-carbonitrile
1-(Bromomethyl)cyclopropane-1-carbonitrile
1-(Bromomethyl)cyclopropane-1-carbonitrile
APOSOR51852-1G
:
Apollo Scientific
:
1-(Bromomethyl)cyclopropane-1-carbonitrile
APOSOR51852-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
160.012 g/mol
:
GHS07
:
Refrigerator
:
98730-78-0
:
C₅H₆BrN
:
MFCD16875700