0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
7-Bromo-2-chloroquinazolin-4-amine
7-Bromo-2-chloroquinazolin-4-amine 95+%
7-Bromo-2-chloroquinazolin-4-amine 95+%
APOSOR500022-250MG
:
Apollo Scientific
:
7-Bromo-2-chloroquinazolin-4-amine 95+%
APOSOR500022-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
258.502 g/mol
:
GHS07
:
Refrigerator
:
1107695-08-8
:
C₈H₅BrClN₃
:
MFCD11858276