Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-5-chloro-4-iodopyridine
2-Bromo-5-chloro-4-iodopyridine
2-Bromo-5-chloro-4-iodopyridine
2-Bromo-5-chloro-4-iodopyridine
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-5-chloro-4-iodopyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
1061357-88-7
:
Formula:
C₅H₂BrClIN
MDL Number:
MFCD16295248
:
318.338 g/mol
Storage Temperature:
Ambient