Order Entry
Sweden
ContactUsLinkComponent
7-Hydroxypyrazolo[1,5-a]pyrimidine-6-carbonitrile
7-Hydroxypyrazolo[1,5-a]pyrimidine-6-carbonitrile
Catalog # APOSOR470558-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Hydroxypyrazolo[1,5-a]pyrimidine-6-carbonitrile
Catalog # APOSOR470558-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.133 g/mol
Storage Temperature:Ambient
CAS Number:89642-01-3
Formula:C₇H₄N₄O
MDL Number:MFCD00973929