Order Entry
Sweden
ContactUsLinkComponent
(1E)-1-(Dimethylamino)-4-(4-methylphenoxy)pent-1-en-3-one, tech
(1E)-1-(Dimethylamino)-4-(4-methylphenoxy)pent-1-en-3-one, tech
Catalog # APOSOR32410-1G
Supplier:  Apollo Scientific
CAS Number:  
(1E)-1-(Dimethylamino)-4-(4-methylphenoxy)pent-1-en-3-one, tech
Catalog # APOSOR32410-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.306 g/mol
Storage Temperature:Ambient
CAS Number:477864-99-6
Formula:C₁₄H₁₉NO₂
MDL Number:MFCD00243598