Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-7-methoxyquinoline-3-methanol
2-Chloro-7-methoxyquinoline-3-methanol
2-Chloro-7-methoxyquinoline-3-methanol
2-Chloro-7-methoxyquinoline-3-methanol
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-7-methoxyquinoline-3-methanol
Supplier:
Apollo Scientific
About this item
CAS Number:
333408-48-3
:
Formula:
C₁₁H₁₀ClNO₂
MDL Number:
MFCD02232331
:
223.656 g/mol
Storage Temperature:
Ambient