Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyclobutane-1,3-diol
Cyclobutane-1,3-diol
Cyclobutane-1,3-diol
APOSOR304355-1G
:
Apollo Scientific
:
Cyclobutane-1,3-diol
APOSOR304355-1G
:
Apollo Scientific
:
Förpackningstyp:
Bottle
Förp.:
1 g
Seq:
3047
:
:
88.105 g/mol
:
Omgivande
:
63518-47-8
:
C₄H₈O₂
:
MFCD22627868