Order Entry
Sweden
ContactUsLinkComponent
 
Irritant
5-Bromo-2-(tetrahydrofuran-3-yl)isoindolin-1-one
Catalog # APOSOR300220-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-2-(tetrahydrofuran-3-yl)isoindolin-1-one
Catalog # APOSOR300220-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :282.133 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:2227272-50-4
Formula:C₁₂H₁₂BrNO₂
MDL Number:MFCD31561252