Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lumiracoxib-acyl-β-D-glucuronide
Lumiracoxib-acyl-β-D-glucuronide ≥93.8%
Lumiracoxib-acyl-β-D-glucuronide ≥93.8%
Lumiracoxib-acyl-β-D-glucuronide ≥93.8%
Supplier:
Apollo Scientific
Certificates
Expand image
Lumiracoxib-acyl-β-D-glucuronide ≥93.8%
Supplier:
Apollo Scientific
About this item
CAS Number:
697287-17-5
:
Storage Temperature:
Ambient