Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-Allose-1,2,3,4,5,6-13C6
D-Allose-1,2,3,4,5,6-13C6 99% min. Isot. 99%
D-Allose-1,2,3,4,5,6-13C6 99% min. Isot. 99%
D-Allose-1,2,3,4,5,6-13C6 99% min. Isot. 99%
Supplier:
Apollo Scientific
Certificates
Expand image
D-Allose-1,2,3,4,5,6-13C6 99% min. Isot. 99%
Supplier:
Apollo Scientific
About this item
:
Formula:
[₁₃C]₆H₁₂O₆
MDL Number:
MFCD28397783
:
186.112 g/mol
Storage Temperature:
Ambient