Lucigenin chemiluminescence reagent, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:2315-97-1
:22103-92-0
Formula:C₂₈H₂₂N₄O₆
MDL Number:MFCD00011925
:510.51 g/mol
Melting Pt:245...250 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour)
Yellow with an Orange to Brown Cast:Conforms - Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- TLC:Consistent with Past Lots
- Solubility (Turbidity)
C = 40 mg/4 ml, Acetic Acid:Clear to Hazy - Solubility (Colour)
Yellow to Very Dark Yellow:Conforms to Requirements