


Papanicolaou's solution modified EA, Sigma-Aldrich®
Catalog # HT40232-1L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
:1