Order Entry
Sweden
ContactUsLinkComponent
 
Lucifer yellow CH dilithium salt for fluorescence Stain, Sigma-Aldrich®
 
undefined
Lucifer yellow CH dilithium salt for fluorescence Stain, Sigma-Aldrich®

 

  • Appearance (Colour):
    Faint Orange to Very Dark Orange
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Solubility (Colour)
    1 mg/ml H2O:
    Faint Yellow to Very Dark Yellow and Faint Orange to Very Dark Orange
  • Solubility (Turbidity):
    Clear to Slightly Hazy
  • Fluorescence:
    Pass