Order Entry
Sweden
ContactUsLinkComponent
 
Solvent Blue 38, Practical grade, Sigma-Aldrich®
 
Solvent Blue 38, Practical grade, Sigma-Aldrich®
 

 

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Nitrogen:
    14.5 - 16.5 %
  • Copper (Cu):
    3.5 - 5.5 %
  • Solubility (Colour)
    1 mg/ml, methanol:
    Dark Blue