

Fast Red Violet LB Salt ≥90% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:32348-81-5
:
Formula:C₁₄H₁₁Cl₂N₃O
MDL Number:MFCD00011892
:M-Clarity™ quality level = MQ100
:376.31 g/mol
:GHS08
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Carbon Content:40.2 - 49.1%
- Nitrogen Content:10.1 - 12.3%
- Dye Content:≥90%
- Solubility (Turbidity), 1 mg/ml, Water:Clear to Slightly Hazy
- Solubility (Colour):Very Faint Yellow to Yellow