illustra™ mRNA purification kit
Catalog # 27-9258-02
Supplier: Cytiva
CAS Number:
Specifications
- Pk:1 KIT
- Description:illustra™ mRNA Purification Kit, 4 purifications (8 columns)
Specifications
About this item
The mRNA Purification Kit is designed for the rapid purification of polyadenylated RNA from eukaryotic total RNA. The kit is based on a specially developed procedure featuring pre-packed spin columns containing oligo(dT)-cellulose.
- Purifies mRNA in 30 to 45 minutes from total RNA extracted from 25 mg to 1 g of cells or tissue
- Features pre-dispensed reagents for each purification, which minimises the risk of nuclease contamination
- Produces mRNA ready for direct use (without precipitation) in procedures such as cDNA synthesis, PCR, Northern blot hybridisation and in vitro translation
- Compatible with total RNA isolated by any currently available method
Delivery: Kit contains oligo(dT)-cellulose columns, glycogen solution, sample buffer, high salt buffer, low salt buffer, elution buffer and protocol. All components are pre-dispensed for each purification.