Order Entry
Sweden
ContactUsLinkComponent
di-Sodium tetraborate, anhydrous ≥98%, AnalaR NORMAPUR®
Health
di-Sodium tetraborate, anhydrous ≥98%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
di-Sodium tetraborate, anhydrous ≥98%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (acidimetric):
    min. 98 %
  • Insoluble in water:
    max. 0,05 %
  • Chlorides (Cl):
    max. 0,05 %
  • Phosphates (as PO4):
    max. 0,005 %
  • Sulfates (SO4):
    max. 0,05 %
  • Calcium (Ca):
    max. 0,02 %
  • Copper (Cu):
    max. 0,005 %
  • Heavy metals (as Pb):
    max. 0,005 %
  • Iron (Fe):
    max. 0,005 %
  • Lead (Pb):
    max. 0,005 %
  • Magnesium (Mg):
    max. 0,02 %
  • Nickel (Ni):
    max. 0,005 %
  • Potassium (K):
    max. 0,05 %

Specification Test Results

Related Information