Order Entry
Sweden
ContactUsLinkComponent
Corning® rat collagen I (from tail tendon)
Corning® rat collagen I (from tail tendon)
Catalog # 734-1097
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
undefined
Corning® rat collagen I (from tail tendon)
Catalog # 734-1097
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Source:
    Tail tendon
  • Species:
    Rat
  • Seq:
    04
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Storage conditions:
    Stable for at least three months at 2 to 8 °C. Do not freeze.
  • Environmental certification:
    ISO 14001 - Bedford
  • Protein/peptide name:
    Collagen I
  • Formulation:
    As a liquid in 0,02 N acetic acid. Typical protein concentration 3 - 4 mg/ml in standard preparation.

Specifications

About this item

Collagen I is found in most tissues and organs, but is most plentiful in dermis, tendon, and bone. Used as a thin coating in cell and tissue culture, it is often used to enhance cell attachment and proliferation. When applied as a gel it can be used to promote expression of cell-specific morphology and function. Collagen I is commonly used to culture endothelial cells, hepatocytes, muscle cells, and a variety of other cell types. The HC formulation of rat tail collagen I is used for three dimensional applications requiring a sturdy gel that provides a maximal 3D support matrix (guidelines are included with product).

Quality control: All preparations are quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma. Source material for human collagen I tested for hepatitis B antigen and HIV-1 antibody.