Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Jack bean Urease
Jack bean Urease
Jack bean Urease
Leverantör:
TCI
Certifikat
Expandera bilden
Jack bean Urease
Leverantör:
TCI
Om denna produkt
Specifikationer
Species:
Jack bean
Enzyme name:
Urease
Specifikationer
Relaterad information
Dokumentation
Manuell
Manual Bioscience and Analyticalscience
Dokumentation