Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Penicillinase
Penicillinase
Penicillinase
Catalog # TCIAP0310-25ML
Supplier:
TCI
CAS Number:
Expand image
Penicillinase
Catalog # TCIAP0310-25ML
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 mL
Enzyme name:
Penicillinase
Specifications
About this item
Product Details
1 ml will inactivate 120000 units of Penicillin
contains 0,25% phenol as preservative substance
Product Details