Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Bovine hyaluronidase (from Testes)
Bovine hyaluronidase (from Testes)
Bovine hyaluronidase (from Testes)
Catalog # TCIAH0164-100MG
Leverantör:
TCI
CAS-nummer:
Expand image
Bovine hyaluronidase (from Testes)
Catalog # TCIAH0164-100MG
Leverantör:
TCI
CAS-nummer:
Specifikationer
Pk:
100 mg
Source:
Testes
Species:
Bovine
Enzyme name:
Hyaluronidase
Specifikationer
Om denna produkt
Product Details
Product Details