Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
1-Butyl-2,3-dimethylimidazolium
1-Butyl-2,3-dimethylimidazolium
1-Butyl-2,3-dimethylimidazolium
Katalog # TCIAB3028-1G
Leverantör:
TCI
CAS-nummer:
Expand image
1-Butyl-2,3-dimethylimidazolium
Katalog # TCIAB3028-1G
Leverantör:
TCI
CAS-nummer:
Specifikationer
Pk:
1 g
Enzyme name:
1-Butyl-2,3-dimethylimidazolium
Specifikationer
Om denna produkt
Product Details
Product Details