Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Aspergillus
genus
Acylase
Aspergillus genus Acylase
Aspergillus genus Acylase
Catalog # TCIAA0688-25G
Supplier:
TCI
CAS Number:
Expand image
Aspergillus genus Acylase
Catalog # TCIAA0688-25G
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 g
Species:
Aspergillus genus
Enzyme name:
Acylase
Specifications
About this item
Product Details
Product Details