Order Entry
Sweden
ContactUsLinkComponent
1,4-Butanesultone
1,4-Butanesultone
Catalog # MOLE40482562-100G
Supplier:  Molekula
CAS Number:  
1,4-Butanesultone
Catalog # MOLE40482562-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :136.17 g/mol
Boiling Pt:153 °C (1013 hPa)
Melting Pt:12.5…15 °C
Melting Pt:12,5…15 °C
Density:1,331 g/cm³ (20 °C)
EINECS:216-647-9
CAS Number:1633-83-6
Formula:C₄H₈O₃S
MDL Number:MFCD00006584