
Tungsten(IV) carbide ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12070-12-1
:
Formula:CW
MDL Number:MFCD00011464
Merck Index:14,9815
Boiling Pt:6000 °C (1013 hPa)
Melting Pt:2870 °C
Density:15.63
EINECS:235-123-0
ADR:4.1,III
:195.85 g/mol
UN:3178
:GHS02
Storage Temperature:Ambient