Order Entry
Sweden
ContactUsLinkComponent
Calcium hypochlorite
Calcium hypochlorite
Catalog # ROTH5164.2
Supplier:  Roth Carl
CAS Number:  
Calcium hypochlorite
Catalog # ROTH5164.2
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications

About this item

 :
 :142.98 g/mol
Merck Index:13,02111
Melting Pt:∼100 °C
Density:2.35 g/cm³ (20 °C)
EINECS:231-908-7
 :GHS03
 :GHS05
 :GHS07
 :GHS09
CAS Number:7778-54-3
Formula:CaCl₂O₂
MDL Number:MFCD00010900