Potassium molybdate(VI), anhydrous ≥99.8% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13446-49-6
:
Formula:K₂MoO₄
MDL Number:MFCD00011370
:238.13 g/mol
Merck Index:14,7647
Melting Pt:919 °C
Density:2.3
EINECS:236-599-2
:GHS07
Storage Temperature:Ambient