Order Entry
Sweden
ContactUsLinkComponent
Water Reag. Ph. Eur. 1095502 carbon dioxide-free
Water Reag. Ph. Eur. 1095502 carbon dioxide-free
Catalog # 87959.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water Reag. Ph. Eur. 1095502 carbon dioxide-free
Catalog # 87959.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332