

trans-1,4-Diaminocyclohexane ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:2615-25-0
:
Formel:C₆H₁₄N₂
MDL-nummer:MFCD00075174
:114.19 g/mol
Kokande Pt:88°C
Smält Pt:70 °C
FN:3259
ADR:8,III
:GHS05
:GHS07
Specifikationstestresultat
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
- Solubility in Methanol:almost transparency