

trans-1,4-Diaminocyclohexane 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2615-25-0
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00075174
:114.19 g/mol
Boiling Pt:199 °C (1013 hPa)
Melting Pt:68…71 °C
Flash Pt:71 °C
Storage Temperature:Ambient
UN:3259
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Appearance:White to yellow to brown Crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:68 to 73 °C
- Water:≤1 %
- GC:≥98.0 %