Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-6-methylphenyl isocyanate
2-Chloro-6-methylphenyl isocyanate
2-Chloro-6-methylphenyl isocyanate
2-Chloro-6-methylphenyl isocyanate
Supplier:
Thermo Scientific
Certificates
Expand image
2-Chloro-6-methylphenyl isocyanate
Supplier:
Thermo Scientific
About this item
CAS Number:
40398-01-4
:
Formula:
C₈H₆ClNO
MDL Number:
MFCD00037030
:
167.59 g/mol
Boiling Pt:
73…75 °C (1 mmHg)
Density:
1,23 g/cm³ (20 °C)