Order Entry
Sweden
ContactUsLinkComponent
Dimethyl-5-nitroisophthalate
Dimethyl-5-nitroisophthalate
Catalog # MOLEM54542162
Supplier:  Molekula
CAS Number:  
Dimethyl-5-nitroisophthalate
Catalog # MOLEM54542162
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :239.18 g/mol
EINECS:236-307-3
CAS Number:13290-96-5
Formula:C₁₀H₉NO₆
MDL Number:MFCD00008429