Order Entry
Sweden
ContactUsLinkComponent
 
Sulpho-KMUS ≥98.0% (by HPLC)
 :  TCI
 
Sulpho-KMUS ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    5.40 to 6.20 %