Sulindac ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:38194-50-2
:
Formel:C₂₀H₁₇FO₃S
MDL-nummer:MFCD00599589
Merck Index:14,08982
ADR:6.1,III
:356.411 g/mol
EINECS:253-819-2
FN:2811
:GHS06
:GHS08
Specifikationstestresultat
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:182.0 to 186.0 deg-C