O-Tritylhydroxylamine ≥98.0% (by HPLC, total nitrogen)
: TCI
:31938-11-1
:
:C₁₉H₁₇NO
:MFCD00042818
:275.35 g/mol
:250-868-1
:GHS07
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:84.0 to 88.0 deg-C