1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:365564-05-2
:
Formula:C₂₄H₃₉B₃O₆
MDL Number:MFCD17167305
:456.00 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %