Tris[4-(5-phenylthiophen-2-yl)phenyl]amine ≥98.0% (by HPLC)
TCIAT3328-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:211.0 to 215.0 deg-C
:
:719.98 g/mol
:803727-09-5
:C₄₈H₃₃NS₃
:MFCD28291947