
2-(Trifluoromethyl)-5-pyridineboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT3188-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
About this item
:
:190.92 g/mol
:GHS07
CAS Number:868662-36-6
Formula:C₆H₅BF₃NO₂
MDL Number:MFCD07375381