(S,S)-Ts-DENEB®
Supplier: TCI
Certificates
About this item
CAS Number:1384974-37-1
:
Formula:C₃₁H₃₃ClN₂O₃RuS
MDL Number:MFCD22666040
:650.20 g/mol
UN:3077
ADR:9,III
:GHS09
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Content (Ruthenium):13 to 17 %