

Trimethylsilyl-2,2-difluoro-2-(fluorosulphonyl)acetate ≥96.0% (by GC)
: TCI
:120801-75-4
:
:C₅H₉F₃O₄SSi
:MFCD02093343
:250.27 g/mol
:86°C
:26°C
:2920
:8,II
:GHS02
:GHS05
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %