Order Entry
Sweden
ContactUsLinkComponent
Tetrakis(dimethylamino)diboron ≥95.0% (by total nitrogen basis)
Tetrakis(dimethylamino)diboron ≥95.0% (by total nitrogen basis)
Catalog # TCIAT3019-1G
Supplier:  TCI
CAS Number:  
undefined
Tetrakis(dimethylamino)diboron ≥95.0% (by total nitrogen basis)
Catalog # TCIAT3019-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (with Total Nitrogen):
    min. 95.0 %

Specification Test Results

About this item

 :
 :197.93 g/mol
Boiling Pt:87°C
Melting Pt:-33°C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1630-79-1
Formula:C₈H₂₄B₂N₄
MDL Number:MFCD00048012
Melting Pt:–33°C