
2-[4-(Trifluoromethyl)phenyl]pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:203065-88-7
:
Formula:C₁₂H₈F₃N
MDL Number:MFCD08235168
:223.20 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:gt. 74.0 to 78.0 deg-C