Triethylene glycol-2-bromoethyl methyl ether ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:110429-45-3
:
Formula:C₉H₁₉BrO₄
MDL Number:MFCD12545963
:271.15 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together