D-ɑ-Tocopherol succinate ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4345-03-3
:
Formula:C₃₃H₅₄O₅
MDL Number:MFCD00072055
:530.79 g/mol
Merck Index:14,09495
Storage Temperature:Kylskåp
EINECS:224-403-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:73.0 to 77.0 deg-C
- Specific rotation [a]20/D:+3.0 to +5.0 deg (C=8, EtOH)